Learn More
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 342613-63-2 |
| Molecular Formula | C9H7BrN2O2 |
| MDL Number | MFCD10566741 |
| Quantity | 250 mg |
| SMILES | O=C(C1=CC2=NC=C(Br)N2C=C1)OC |
| Molecular Weight (g/mol) | 255.07 |
| Percent Purity | 95% |
| Chemical Name or Material | Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate |