Läs mer
Methyl 3-hydroxy-4,5-dimethoxybenzoate, TRC
Produktkod. 30483347
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30483347 2.5 g 2.5g
30473240 500 mg -
30469846 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7805052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83011-43-2
Molekylformel C10H12O5
Molekylvikt (g/mol) 212.2
Synonym Methyl 4,5-dimethoxy-3-hydroxybenzoate
IUPAC-namn methyl 3-hydroxy-4,5-dimethoxybenzoate
LEDER COC(=O)c1cc(O)c(OC)c(OC)c1
Specifikationer
Kemiskt namn eller material Methyl 3-hydroxy-4,5-dimethoxybenzoate
Kvantitet 2.5 g
InChI formel InChI=1S/C10H12O5/c1-13-8-5-6(10(12)15-3)4-7(11)9(8)14-2/h4-5,11H,1-3H3
Formel vikt 212.0685