Läs mer
Methyl 3-Hydroxyanthranilate, TRC
Produktkod. 30468022
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30468022 250 mg
1 options
Produktkod. 30468022 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM316850250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-Hydroxyanthranilate
CAS 17672-21-8
Molekylformel C8H9NO3
Kvantitet 250 mg
Synonym 2-Amino-3-hydroxybenzoic Acid Methyl Ester, Methyl 2-Amino-3-hydroxybenzoate, 3-Hydroxyanthranilic Acid Methyl Ester, 2-Amino-3-hydroxybenzoic Acid Methyl Ester
InChI formel InChI=1S/C8H9NO3/c1-12-8(11)5-3-2-4-6(10)7(5)9/h2-4,10H,9H2,1H3
LEDER COC(=O)c1cccc(O)c1N
IUPAC-namn methyl 2-amino-3-hydroxybenzoate
Molekylvikt (g/mol) 167.16
Formel vikt 167.0582