Läs mer
Methyl 3-Mercaptobenzoate, ~85%, TRC
Produktkod. 30464496
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464496 5 g
30464716 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30464496 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3207305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72886-42-1
Molekylformel C8 H8 O2 S
Molekylvikt (g/mol) 168.21
IUPAC-namn methyl 3-sulfanylbenzoate
LEDER COC(=O)c1cccc(S)c1
Specifikationer
Kemiskt namn eller material Methyl 3-Mercaptobenzoate
Kvantitet 5 g
InChI formel InChI=1S/C8H8O2S/c1-10-8(9)6-3-2-4-7(11)5-6/h2-5,11H,1H3
Formel vikt 168.0245