Learn More
Methyl 3-mercaptopropionate, 98%, Acros Organics
201.66 SEK - 1691.55 SEK
Chemical Identifiers
CAS 2935-90-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00004895
InChI Key LDTLDBDUBGAEDT-UHFFFAOYSA-N
Synonym methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
PubChem CID 18050
IUPAC Name methyl 3-sulfanylpropanoate
SMILES COC(=O)CCS
View More Specs
Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10532351 SDS View Documents Acros Organics
148870050
5g Glass bottle 201.66 SEK
5g View alternative products
Save Up to 
»
EU Stock 5
Add to basket
10775031 SDS View Documents Acros Organics
148871000
100g Glass bottle 509.50 SEK
100g View alternative products
Save Up to 
»
EU Stock 2
Add to basket
10021510 SDS View Documents Acros Organics
148875000
500g Glass bottle 1691.55 SEK
500g View alternative products
Save Up to 
»
EU Stock 2
Add to basket
Specifications
Chemical Identifiers
2935-90-2
120.17
LDTLDBDUBGAEDT-UHFFFAOYSA-N
18050
COC(=O)CCS
C4H8O2S
MFCD00004895
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
methyl 3-sulfanylpropanoate
Specifications
Methyl 3-mercaptopropionate
60°C
Glass bottle
1.08
Colorless
98%
98%
HSCH2CH2CO2CH3
03, II, 214
LDTLDBDUBGAEDT-UHFFFAOYSA-N
methyl 3-sulfanylpropanoate
18050
Liquid
1.0800g/mL
Authentic
1.4620 to 1.4640
166.0°C to 169.0°C
5g
2935-90-2
C4H8O2S
MFCD00004895
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
COC(=O)CCS
120.17
120.17
98%
Safety and Handling
Safety and Handling
Product Identifier
  • Methyl 3-mercaptopropionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.