Learn More
Methyl 3-methoxythiophene-2-carboxylate, 97%
Product Code. 11458987
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458987 5 g 5g
11448987 1 g 1g
11468987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11458987 Supplier Thermo Scientific Alfa Aesar Supplier No. B20586.06
 Restricted Item
Beräknad leverans: 25-11-2026
Add to basket
CAS: 62353-75-7 | C7H8O3S | 172.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62353-75-7
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00102115
InChI Key MWADWXJPBMFRGL-UHFFFAOYSA-N
Synonym 3-methoxy-2-thiophenecarboxylic acid methyl ester, acmc-20abea, methyl 3-methoxythiophene carboxylate, methyl 3-methoxy-2-thiophenecarboxylate, 2-thiophenecarboxylicacid,3-methoxy-,methyl ester, 3-methoxythiophene-2-carboxylic acid methyl ester
PubChem CID 701637
IUPAC Name methyl 3-methoxythiophene-2-carboxylate
SMILES COC(=O)C1=C(OC)C=CS1
Specifications
Melting Point 60°C to 63°C
Quantity 5 g
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material Methyl 3-methoxythiophene-2-carboxylate