Läs mer
Methyl 3-(Methylthio)benzoate, TRC
Produktkod. 30468305
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30468305 500 mg
1 options
Produktkod. 30468305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM238635500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-(Methylthio)benzoate
CAS 90721-40-7
Molekylformel C9H10O2S
Kvantitet 500 mg
Synonym 3-Methylsulfanylbenzoic acid methyl ester, Methyl m-(methoxycarbonyl)phenyl sulfide, Methyl m-(methylthio)benzoate
InChI formel InChI=1S/C9H10O2S/c1-11-9(10)7-4-3-5-8(6-7)12-2/h3-6H,1-2H3
LEDER COC(=O)c1cccc(SC)c1
IUPAC-namn methyl 3-methylsulfanylbenzoate
Molekylvikt (g/mol) 182.24
Formel vikt 182.0402