Läs mer
Methyl 3-Oxododecanoate, TRC
Produktkod. 30475158
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30475158 1 g
1 options
Produktkod. 30475158 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3247201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-Oxododecanoate
CAS 76835-64-8
Molekylformel C13H24O3
Kvantitet 1 g
Synonym Methyl Dodecanon-3-oate, Methyl β-Oxododecanoate, 3-Oxo-dodecanoic Acid Methyl Ester
InChI formel InChI=1S/C13H24O3/c1-3-4-5-6-7-8-9-10-12(14)11-13(15)16-2/h3-11H2,1-2H3
LEDER O=C(CC(OC)=O)CCCCCCCCC
IUPAC-namn methyl 3-oxododecanoate
Molekylvikt (g/mol) 228.33
Formel vikt 228.1725