Learn More
Methyl 3-Oxopentanoate, Sigma-Aldrich™
Product Code. 15840441
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity Unit Size
15840441 10 mL 10mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15840441 Supplier Sigma Aldrich Supplier No. 8184920010
 Restricted Item
Beräknad leverans: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 30414-53-0
Density 1.04 g/cm3 at 25°C
MDL Number MFCD00011705
Quantity 10 mL
Synonym Methyl 3-oxopentanoate, 3-Oxopentanoic acid methyl ester, Methyl 3-oxopentanoate
Molecular Weight (g/mol) 130.14
Grade Research