Läs mer
Methyl 3-Oxotetradecanoate, TRC
Produktkod. 30467488
Change view
Kvantitet:
500 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30467488 1 g -
30479483 500 mg -
30481055 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467488 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3258651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22348-97-6
Molekylformel C15 H28 O3
Molekylvikt (g/mol) 256.38
IUPAC-namn methyl 3-oxotetradecanoate
LEDER CCCCCCCCCCCC(=O)CC(=O)OC
Specifikationer
Kemiskt namn eller material Methyl 3-Oxotetradecanoate
Kvantitet 1 g
InChI formel InChI=1S/C15H28O3/c1-3-4-5-6-7-8-9-10-11-12-14(16)13-15(17)18-2/h3-13H2,1-2H3
Formel vikt 256.2038