Läs mer
Methyl 3-[t-Butyloxycarbonyl)amino]-L-alanine, TRC
Produktkod. 30479281
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30479281 100 mg
1 options
Produktkod. 30479281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-[(t-Butyloxycarbonyl)amino]-L-alanine
CAS 77087-60-6
Molekylformel C9H18N2O4
Kvantitet 100 mg
Synonym 3-[[(1,1-Dimethylethoxy)carbonyl]amino]-L-alanine Ethyl Ester, (2S)-2-Amino-3-[(tert-butoxycarbonyl)amino]propionic Acid Methyl Ester
InChI formel InChI=1S/C9H18N2O4/c1-9(2,3)15-8(13)11-5-6(10)7(12)14-4/h6H,5,10H2,1-4H3,(H,11,13)/t6-/m0/s1
LEDER COC(=O)[C@@H](N)CNC(=O)OC(C)(C)C
IUPAC-namn methyl (2S)-2-amino-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Molekylvikt (g/mol) 218.25
Formel vikt 218.1267