Läs mer
Methyl (3S)-3-Hydroxyhexadecanoate, TRC
Produktkod. 30465773
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30465773 250 mg
1 options
Produktkod. 30465773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM312778250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (3S)-3-Hydroxyhexadecanoate
CAS 76898-45-8
Molekylformel C17H34O3
Kvantitet 250 mg
Synonym (3S)-3-Hydroxy-hexadecanoic Acid Methyl Ester, (S)-3-Hydroxy-hexadecanoic Acid Methyl Ester
InChI formel InChI=1S/C17H34O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-16(18)15-17(19)20-2/h16,18H,3-15H2,1-2H3/t16-/m0/s1
LEDER CCCCCCCCCCCCC[C@H](O)CC(=O)OC
IUPAC-namn methyl (3S)-3-hydroxyhexadecanoate
Molekylvikt (g/mol) 286.45
Formel vikt 286.2508