Läs mer
Methyl 4-(2,4-Dichlorobenzoyl)-1H-pyrrole-2-carboxylate, TRC
Produktkod. 30471963
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471963 1 g
30481376 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471963 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1379801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-(2,4-Dichlorobenzoyl)-1H-pyrrole-2-carboxylate
CAS 453557-85-2
Molekylformel C13H9Cl2NO3
Kvantitet 1 g
InChI formel InChI=1S/C13H9Cl2NO3/c1-19-13(18)11-4-7(6-16-11)12(17)9-3-2-8(14)5-10(9)15/h2-6,16H,1H3
LEDER COC(=O)c1cc(c[nH]1)C(=O)c2ccc(Cl)cc2Cl
IUPAC-namn methyl 4-(2,4-dichlorobenzoyl)-1H-pyrrole-2-carboxylate
Molekylvikt (g/mol) 298.12
Formel vikt 296.9959