Läs mer
Methyl 4-(4-(3-(4-((2-(Methylcarbamoyl)pyridin-4-yl)oxy)phenyl)ureido)phenoxy)picolinate, TRC
Produktkod. 30366949
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30366949 50 mg
1 options
Produktkod. 30366949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29709550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-(4-((4-((2-(Methylcarbamoyl)-4-pyridyl)oxy)phenyl)carbamoylamino)phenoxy)pyridine-2-carboxylate
CAS 2204442-52-2
Molekylformel C27 H23 N5 O6
Kvantitet 50 mg
Synonym Des-(4-chloro-3-trifluoromethyl)-4-(methyl-4-hydroxypicolinate) Sorafenib, 2-Pyridinecarboxylic acid, 4-[4-[[[[4-[[2-[(methylamino)carbonyl]-4-pyridinyl]oxy]phenyl]amino]carbonyl]amino]phenoxy]-, methyl ester (ACI), Methyl 4-[4-[[4-[[2-(methylcarbamoyl)-4-pyridyl]oxy]phenyl]carbamoylamino]phenoxy]pyridine-2-carboxylate
InChI formel InChI=1S/C27H23N5O6/c1-28-25(33)23-15-21(11-13-29-23)37-19-7-3-17(4-8-19)31-27(35)32-18-5-9-20(10-6-18)38-22-12-14-30-24(16-22)26(34)36-2/h3-16H,1-2H3,(H,28,33)(H2,31,32,35)
LEDER CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Oc4ccnc(c4)C(=O)OC)cc3)cc2)ccn1
IUPAC-namn methyl 4-[4-[[4-[2-(methylcarbamoyl)pyridin-4-yl]oxyphenyl]carbamoylamino]phenoxy]pyridine-2-carboxylate
Molekylvikt (g/mol) 513.501
Formel vikt 513.165