Läs mer
Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-(methylsulfonyl)pyrimidine-5-carboxylate, TRC
Produktkod. 30465008
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30465008 10 mg
1 options
Produktkod. 30465008 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30392010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-(methylsulfonyl)pyrimidine-5-carboxylate
CAS 799842-06-1
Molekylformel C16H17FN2O4S
Kvantitet 10 mg
Synonym 5-Pyrimidinecarboxylic acid','4-(4-fluorophenyl)-6-(1-methylethyl)-2-(methylsulfonyl)-','methyl ester, Methyl 4-(4-fluorophenyl)-6-(1-methylethyl)-2-(methylsulfonyl)-5-pyrimidinecarboxylate, 4-(4-Fluorophenyl)-6-isopropyl-2-(methanesulfonyl)pyrimidine-5-carboxylic acid methyl ester, Methyl 4-(4-fluorophenyl)-6-isopropyl-2-(methylsulfonyl)pyrimidine-5-carboxylate
InChI formel InChI=1S/C16H17FN2O4S/c1-9(2)13-12(15(20)23-3)14(10-5-7-11(17)8-6-10)19-16(18-13)24(4,21)22/h5-9H,1-4H3
LEDER COC(=O)c1c(nc(nc1c2ccc(F)cc2)S(=O)(=O)C)C(C)C
IUPAC-namn methyl 4-(4-fluorophenyl)-2-methylsulfonyl-6-propan-2-ylpyrimidine-5-carboxylate
Molekylvikt (g/mol) 352.38
Formel vikt 352.0893