Läs mer
Methyl 4-[(4-Methylpiperazin-1-yl)methyl]benzoate, Mikromol
Produktkod. 30458185
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458185 4 x 25 mg
30050980 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30458185 Leverantör Mikromol Leverantörsnummer MM3566.0700254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-[(4-Methylpiperazin-1-yl)methyl]benzoate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-[(4-Methylpiperazin-1-yl)methyl]benzoate
CAS 314268-40-1
Analyt- eller komponentnamn Methyl 4-[(4-Methylpiperazin-1-yl)methyl]benzoate
Frakt skick Room Temperature
Molekylformel C14 H20 N2 O2
Certifikat och efterlevnad ISO 9001
Synonym Benzoic acid, 4-[(4-methyl-1-piperazinyl)methyl]-, methyl ester, 4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid methyl ester, 4-[(4-Methylpiperazino)methyl]benzoic acid methyl ester, Methyl 4-((4-methylpiperazin-1-yl)methyl)benzoate, Methyl 4-[(4-methylpiperazino)methyl]benzoate
InChI formel InChI=1S/C14H20N2O2/c1-15-7-9-16(10-8-15)11-12-3-5-13(6-4-12)14(17)18-2/h3-6H,7-11H2,1-2H3
Lösningstyp Neat
LEDER COC(=O)c1ccc(CN2CCN(C)CC2)cc1
IUPAC-namn methyl 4-[(4-methylpiperazin-1-yl)methyl]benzoate
Molekylvikt (g/mol) 248.32
Kvantitet 4 x 25 mg
Formel vikt 248.1525 g/mol