missing translation for 'onlineSavingsMsg'
Läs mer

Glassy carbon spherical powder, 200-400 micron, type 1

Produktkod. 11369797
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
50 g
250 g
Förpackningsstorlek:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11369797 250 g 250g
11379797 10 g 10g
11359797 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11369797 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 042130.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

Glassy carbon spherical powder is used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

MDL-nummer MFCD00133992

Specifikationer

Kemiskt namn eller material Glassy carbon spherical powder
Förpackning Plastic bottle
Kvantitet 250 g
UN-nummer UN1325
Löslighetsinformation Insoluble in water.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.