missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,6-dichloronicotinate, 95%, Thermo Scientific Chemicals

Catalog Number p-7055486
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
65973-52-6
C7H5Cl2NO2
206.02
MFCD04125732
JEJMDUMJSZTJTI-UHFFFAOYSA-N
2764083
methyl 4,6-dichloropyridine-3-carboxylate
COC(=O)C1=CN=C(Cl)C=C1Cl
This item is not returnable. View return policy
65973-52-6
C7H5Cl2NO2
206.02
MFCD04125732
JEJMDUMJSZTJTI-UHFFFAOYSA-N
2764083
methyl 4,6-dichloropyridine-3-carboxylate
COC(=O)C1=CN=C(Cl)C=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 13279782
Synonym methyl 4,6-dichloronicotinate, methyl4,6-dichloronicotinate, 4,6-dichloro-nicotinic acid methyl, 4,6-dichloro-nicotinic acid methyl ester, 4,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, methyl ester, 2-p-tolylimidazo 1,2-a pyridine, pubchem12979, ksc494c6d, methyl-4,6-dichloronicotinate
Formula Weight 206.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 4,6-dichloronicotinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'