Läs mer
Methyl 4-Acetamido-5-chloro-2-ethoxybenzoate, TRC
Produktkod. 30474580
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474580 100 mg
30476152 50 mg
30483757 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474580 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM293263100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4235-43-2
Molekylformel C12H14ClNO4
Molekylvikt (g/mol) 271.7
Synonym Methyl 4-(Acetylamino)-5-chloro-2-ethoxybenzoate
IUPAC-namn methyl 4-acetamido-5-chloro-2-ethoxybenzoate
LEDER CCOc1cc(NC(=O)C)c(Cl)cc1C(=O)OC
Specifikationer
Kemiskt namn eller material Methyl 4-Acetamido-5-chloro-2-ethoxybenzoate
Kvantitet 100 mg
InChI formel InChI=1S/C12H14ClNO4/c1-4-18-11-6-10(14-7(2)15)9(13)5-8(11)12(16)17-3/h5-6H,4H2,1-3H3,(H,14,15)
Formel vikt 271.0611