missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7053580
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
18595-14-7
C9H11NO2
165.19
MFCD00102230
ZHIPSMIKSRYZFV-UHFFFAOYSA-N
4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate
2736799
methyl 4-amino-3-methylbenzoate
COC(=O)C1=CC=C(N)C(C)=C1
This item is not returnable. View return policy
18595-14-7
C9H11NO2
165.19
MFCD00102230
ZHIPSMIKSRYZFV-UHFFFAOYSA-N
4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate
2736799
methyl 4-amino-3-methylbenzoate
COC(=O)C1=CC=C(N)C(C)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 116°C to 120°C
Quantity 25 g
Formula Weight 165.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-amino-3-methylbenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'