Läs mer
Methyl 4-Aza-3-oxo-androstane-17Beta-carboxylate, TRC
Produktkod. 30478965
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30478965 50 mg
1 options
Produktkod. 30478965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28858550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Aza-3-oxo-androstane-17beta-carboxylate
CAS 73671-92-8
Molekylformel C20H31NO3
Kvantitet 50 mg
Synonym Methyl 3-Oxo-4-aza-5alpha-androstane-17beta-carboxylate, Methyl (5alpha,17beta)-3-Oxo-4-aza-5alpha-androstane-17-carboxylate, 4-Aza-3-oxo-androstane-17beta-carboxylic Acid Methyl Ester, 3-Oxo-4-aza-5alpha-androstane-17beta-carboxylic Acid Methyl Ester, 3-Oxo-4-aza-5alpha-androstan-17beta-carboxylic Acid Methyl Ester, [4aR-(4aalpha,4bbeta,6aalpha,7alpha,9abeta,9balpha,11abeta)]-1H-Indeno[5,4-f]quinoline-7-carboxylic Acid, hexadecahydro-4a,6a-dimethyl-2-oxo-methyl Ester
InChI formel InChI=1S/C20H31NO3/c1-19-10-8-14-12(13(19)5-6-15(19)18(23)24-3)4-7-16-20(14,2)11-9-17(22)21-16/h12-16H,4-11H2,1-3H3,(H,21,22)/t12-,13-,14-,15+,16+,19-,20+/m0/s1
LEDER COC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn methyl (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxylate
Molekylvikt (g/mol) 333.47
Formel vikt 333.2304