Läs mer
Methyl 4-Bromomethylcinnamate, TRC
Produktkod. 30471987
Change view
Kvantitet:
2 g
Produktkod. Kvantitet
30471987 2 g
1 options
Produktkod. 30471987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2942902G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Bromomethylcinnamate
CAS 946-99-6
Molekylformel C11H11BrO2
Kvantitet 2 g
Synonym 3-[4-(Bromomethyl)phenyl]-2-propenoic Acid Methyl Ester, p-(Bromomethyl)cinnamic Acid Methyl Ester, 3-(4-Bromomethylphenyl)acrylic Acid Methyl Ester
InChI formel InChI=1S/C11H11BrO2/c1-14-11(13)7-6-9-2-4-10(8-12)5-3-9/h2-7H,8H2,1H3/b7-6+
LEDER COC(=O)\C=C\c1ccc(CBr)cc1
IUPAC-namn methyl (E)-3-[4-(bromomethyl)phenyl]prop-2-enoate
Molekylvikt (g/mol) 255.11
Formel vikt 253.9942