Läs mer
Methyl 4-(Butylamino)benzoate, TRC
Produktkod. 30240942
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240942 100 mg
30240489 50 mg
30240553 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM275160100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71839-12-8
Molekylformel C12 H17 N O2
Molekylvikt (g/mol) 207.27
Synonym Methyl 4-(Butylamino)benzoate, Tetracaine Hydrochloride Imp. C (EP)
IUPAC-namn methyl 4-(butylamino)benzoate
LEDER CCCCNc1ccc(cc1)C(=O)OC
Specifikationer
Kemiskt namn eller material Methyl 4-(Butylamino)benzoate
Kvantitet 100 mg
InChI formel InChI=1S/C12H17NO2/c1-3-4-9-13-11-7-5-10(6-8-11)12(14)15-2/h5-8,13H,3-4,9H2,1-2H3
Formel vikt 207.1259