Läs mer
Methyl-4-Chloro-7-methoxyquinoline-6-carboxylate, TRC
Produktkod. 30475098
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30475098 250 mg
1 options
Produktkod. 30475098 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294465250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl-4-Chloro-7-methoxyquinoline-6-carboxylate
CAS 205448-66-4
Molekylformel C12H10ClNO3
Kvantitet 250 mg
Synonym 4-Chloro-7-methoxyquinoline-6-carboxylic Acid Methyl Ester, 4-Chloro-7-methoxy-6-quinolinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C12H10ClNO3/c1-16-11-6-10-7(9(13)3-4-14-10)5-8(11)12(15)17-2/h3-6H,1-2H3
LEDER COC(=O)c1cc2c(Cl)ccnc2cc1OC
Molekylvikt (g/mol) 251.67
Formel vikt 251.0349