Läs mer
Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, TRC
Produktkod. 30474140
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30474140 1 g
1 options
Produktkod. 30474140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3128451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
CAS 35511-15-0
Molekylformel C11 H11 N O5 S
Kvantitet 1 g
Synonym Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Meloxicam Imp. E (EP), Piroxicam Imp. J (EP), 2H-1,2-Benzothiazine-3-carboxylic acid','4-hydroxy-2-methyl-','methyl ester','1,1-dioxide, 2-Methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylic acid methyl ester 1,1-dioxide, 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylic acid methyl ester 1,1-dioxide
InChI formel InChI=1S/C11H11NO5S/c1-12-9(11(14)17-2)10(13)7-5-3-4-6-8(7)18(12,15)16/h3-6,13H,1-2H3
LEDER COC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C
IUPAC-namn methyl 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxylate
Molekylvikt (g/mol) 269.27
Formel vikt 269.0358