Läs mer
Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Produktkod. 30461993
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461993 4 x 25 mg
17961946 25 mg
17922386 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461993 Leverantör Mikromol Leverantörsnummer MM0084.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
CAS 35511-14-9
Analyt- eller komponentnamn Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
Frakt skick Room Temperature
Molekylformel C10 H9 N O5 S
Certifikat och efterlevnad ISO 17025
Synonym Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Piroxicam Imp. G (EP), Piroxicam Impurity G
InChI formel InChI=1S/C10H9NO5S/c1-16-10(13)8-9(12)6-4-2-3-5-7(6)17(14,15)11-8/h2-5,11-12H,1H3
Lösningstyp Neat
LEDER COC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1
IUPAC-namn methyl 4-hydroxy-1,1-dioxo-2H-1λ^{6},2-benzothiazine-3-carboxylate
Molekylvikt (g/mol) 255.25
Kvantitet 4 x 25 mg
Formel vikt 255.0201 g/mol