Läs mer
Methyl 4-Hydroxy Cinnamate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl 4-Hydroxy Cinnamate |
| CAS | 19367-38-5 |
| Molekylformel | C10 H10 O3 |
| Kvantitet | 1 g |
| Synonym | 2-Propenoic acid','3-(4-hydroxyphenyl)-','methyl ester','(2E)-, 2-Propenoic acid','3-(4-hydroxyphenyl)-','methyl ester','(E)-, Cinnamic acid','p-hydroxy-','methyl ester','(E)- (8CI), (E)-3-(4-Hydroxyphenyl)acrylic acid methyl ester, (E)-4-Hydroxycinnamic acid methyl ester, (E)-Methyl 3-(4-hydroxyphenyl)acrylate, (E)-Methyl p-hydroxycinnamate, Methyl (2E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (2E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (E)-3-(4-hydroxyphenyl)-2-propenoate, Methyl (E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (E)-4-hydroxycinnamate, Methyl (E)-p-coumarate, Methyl (E)-p-hydroxy-cinnamate, Methyl (E)-p-hydroxycinnamate, Methyl p-hydroxy-(E)cinnamate, Methyl p-hydroxy-trans-cinnamate, Methyl trans-4-hydroxycinnamate, Methyl trans-p-coumarate, trans-Methyl p-hydroxycinnamate, trans-p-Coumaric acid methyl ester, trans-p-Hydroxycinnamic acid methyl ester |
| InChI formel | InChI=1S/C10H10O3/c1-13-10(12)7-4-8-2-5-9(11)6-3-8/h2-7,11H,1H3/b7-4+ |
| LEDER | COC(=O)\C=C\c1ccc(O)cc1 |
| IUPAC-namn | methyl (E)-3-(4-hydroxyphenyl)prop-2-enoate |
| Molekylvikt (g/mol) | 178.18 |
| Formel vikt | 178.063 |