missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Methyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Specifications
Specifications
| Kemiskt namn eller material | Methyl Paraben-d4 |
| CAS | 362049-51-2 |
| Molekylformel | C8 2H4 H4 O3 |
| Kvantitet | 0.05 g |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, methyl ester (9 CI), Methyl 4-hydroxybenzoate-2,3,5,6-D4, Methyl 4-hydroxybenzoate-d4, Methyl parahydroxybenzoate-d4, 4-Hydroxybenzoic-2,3,5,6-d4 acid methyl ester, Methylparaben-d4 |
| InChI formel | InChI=1 S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9 H,1H3/i2D,3 D,4 D,5 D |
| LEDER | [2 H]c1c([2 H])c(C(=O)OC)c([2 H])c([2 H])c1O |
| IUPAC-namn | methyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate |
| Molekylvikt (g/mol) | 156.17 |
| Formel vikt | 156.0725 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?