Läs mer
Methyl 4-Methoxyacetoacetate, TRC
Produktkod. 30474336
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30474336 10 g
1 options
Produktkod. 30474336 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32084510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Methoxyacetoacetate
CAS 41051-15-4
Molekylformel C35 H32 Cl N3 O3 S
Kvantitet 10 g
Synonym 4-Methoxy-3-oxo-butyric Acid Methyl ester, 4-Methoxy-3-oxobutanoic Acid Methyl Ester, 4-Methoxyacetoacetic Acid Methyl Ester, Methyl 4-Methoxy-3-oxobutanoate, Methyl 4-Methoxy-3-oxobutyrate, Methyl 4-Methoxyacetoacetate, Methyl gamma-Methoxyacetoacetate, gamma-Methoxyacetoacetic Acid Methyl Ester
InChI formel InChI=1S/C35H32ClN3O3S/c1-34(2,3)32(41)42-23-24-14-13-21-28(36)30(24)38-31(40)29-22-37-33(43-29)39-35(25-15-7-4-8-16-25,26-17-9-5-10-18-26)27-19-11-6-12-20-27/h4-22H,23H2,1-3H3,(H,37,39)(H,38,40)
LEDER CC(C)(C)C(=O)OCc1cccc(Cl)c1NC(=O)c2cnc(NC(c3ccccc3)(c4ccccc4)c5ccccc5)s2
IUPAC-namn [3-chloro-2-[[2-(tritylamino)-1,3-thiazole-5-carbonyl]amino]phenyl]methyl 2,2-dimethylpropanoate
Molekylvikt (g/mol) 610.165
Formel vikt 609.185