Läs mer
Methyl 4-Methyl-3-hydroxybenzoate, TRC
Produktkod. 30478306
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30478306 2.5 g
1 options
Produktkod. 30478306 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3229702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Methyl-3-hydroxybenzoate
CAS 3556-86-3
Molekylformel C9H10O3
Kvantitet 2.5 g
Synonym 3-Hydroxy-4-methylbenzoic Acid Methyl Ester, Methyl 3-Hydroxy-4-methylbenzoate, NSC 604566
InChI formel InChI=1S/C9H10O3/c1-6-3-4-7(5-8(6)10)9(11)12-2/h3-5,10H,1-2H3
LEDER COC(=O)c1ccc(C)c(O)c1
IUPAC-namn methyl 3-hydroxy-4-methylbenzoate
Molekylvikt (g/mol) 166.17
Formel vikt 166.063