Läs mer
Methyl(4-methylnaphthalen-1-ylmethyl)amine, TRC
Produktkod. 30469138
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469138 50 mg
30469139 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469138 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32318550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98978-50-8
Molekylformel C13H15N
Molekylvikt (g/mol) 185.26
Synonym N,4-Dimethyl-1-naphthalenemethanamine
IUPAC-namn N-methyl-1-(4-methylnaphthalen-1-yl)methanamine
LEDER CC1=C2C=CC=CC2=C(CNC)C=C1
Specifikationer
Kemiskt namn eller material Methyl(4-methylnaphthalen-1-ylmethyl)amine
Kvantitet 50 mg
InChI formel InChI=1S/C13H15N/c1-10-7-8-11(9-14-2)13-6-4-3-5-12(10)13/h3-8,14H,9H2,1-2H3
Formel vikt 185.1204