Läs mer
Methyl 4-Nitrophenoxyacetate, TRC
Produktkod. 30468116
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30468116 1 g
1 options
Produktkod. 30468116 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3244831G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 4-Nitrophenoxyacetate
CAS 19786-48-2
Molekylformel C9H9NO5
Kvantitet 1 g
Synonym Methyl 2-(4-Nitrophenoxy)acetate, Methyl p-Nitrophenoxyacetate, (4-Nitrophenoxy)acetic Acid Methyl Ester, (p-Nitrophenoxy)-acetic Acid Methyl Ester, (4-Nitrophenoxy)-acetic Acid Methyl Ester
InChI formel InChI=1S/C9H9NO5/c1-14-9(11)6-15-8-4-2-7(3-5-8)10(12)13/h2-5H,6H2,1H3
LEDER COC(=O)COc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn methyl 2-(4-nitrophenoxy)acetate
Molekylvikt (g/mol) 211.17
Formel vikt 211.0481