missing translation for 'onlineSavingsMsg'
Läs mer

Methyl 4-Oxocyclohexanecarboxylate, TRC

Produktkod. 30475461
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475461 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30475461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM315045250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Methyl 4-Oxocyclohexanecarboxylate
CAS 6297-22-9
Molekylformel C8H12O3
Kvantitet 250 mg
Synonym 4-oxo-Cyclohexanecarboxylic Acid Methyl Ester, 4-(Methoxycarbonyl)cyclohexanone, Methyl 4-Ketocyclohexanecarboxylate, Methyl cyclohexanone-4-carboxylate, NSC 17321
InChI formel InChI=1S/C8H12O3/c1-11-8(10)6-2-4-7(9)5-3-6/h6H,2-5H2,1H3
LEDER COC(=O)C1CCC(=O)CC1
IUPAC-namn methyl 4-oxocyclohexane-1-carboxylate
Molekylvikt (g/mol) 156.18
Formel vikt 156.0786
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.