Läs mer
Methyl 5-Aminopentanoate Hydrochloride, TRC
Produktkod. 30474936
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30474936 50 mg
1 options
Produktkod. 30474936 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28705050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 5-Aminopentanoate Hydrochloride
CAS 29840-56-0
Molekylformel C6H13NO2 . (HCl)
Kvantitet 50 mg
Synonym Methyl 5-Aminovalerate Hydrochloride
InChI formel InChI=1S/C6H13NO2.ClH/c1-9-6(8)4-2-3-5-7;/h2-5,7H2,1H3;1H
LEDER Cl.COC(=O)CCCCN
IUPAC-namn methyl 5-aminopentanoate;hydrochloride
Molekylvikt (g/mol) 167.63
Formel vikt 167.0713