missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-furoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7077245
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2527-99-3
C6H5BrO3
205.01
FBPIDMAELBIRLE-UHFFFAOYSA-N
methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
599633
methyl 5-bromofuran-2-carboxylate
COC(=O)C1=CC=C(O1)Br
This item is not returnable. View return policy
2527-99-3
C6H5BrO3
205.01
FBPIDMAELBIRLE-UHFFFAOYSA-N
methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
599633
methyl 5-bromofuran-2-carboxylate
COC(=O)C1=CC=C(O1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65.0°C to 67.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 205.01
Percent Purity 98%
Chemical Name or Material Methyl 5-bromo-2-furoate
compliance-icons
Product Identifier
  • Methyl 5-bromo-2-furoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'