missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 5-bromo-3-methylpyridine-2-carboxylate, 95%

Brand:  Alfa Aesar™ H64151.06

Additional Details : CAS Number : 213771-32-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15471219

  • 3702.33 SEK / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 5-bromo-3-methylpyridine-2-carboxylate
230.06
5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester
213771-32-5
MFCD07375126

Specifications

Methyl 5-bromo-3-methylpyridine-2-carboxylate
Solid
230.06
5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester
213771-32-5
95%
MFCD07375126
5g
Videos
SDS