missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-4-chloro-2-fluorobenzoate, TRC

Product Code. 30466124
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30466124 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466124 Supplier Toronto Research Chemicals Supplier No. TRCM295855100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methyl 5-bromo-4-chloro-2-fluorobenzoate
CAS 951884-02-9
Molekylformel C8H5BrClFO2
Kvantitet 100 mg
InChI formel InChI=1S/C8H5BrClFO2/c1-13-8(12)4-2-5(9)6(10)3-7(4)11/h2-3H,1H3
LEDER COC(=O)c1cc(Br)c(Cl)cc1F
IUPAC-namn methyl 5-bromo-4-chloro-2-fluorobenzoate
Molekylvikt (g/mol) 267.48
Formel vikt 265.9145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.