Läs mer
Methyl 5-Bromo-4-methoxypicolinate, TRC
Produktkod. 30483514
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483514 5 g
30475097 1 g
30481418 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483514 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2968035G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1256789-95-3
Molekylformel C8H8NO3Br
Molekylvikt (g/mol) 246.05
IUPAC-namn methyl 5-bromo-4-methoxypyridine-2-carboxylate
LEDER COC(=O)c1cc(OC)c(Br)cn1
Specifikationer
Kemiskt namn eller material Methyl 5-Bromo-4-methoxypicolinate
Kvantitet 5 g
InChI formel InChI=1S/C8H8BrNO3/c1-12-7-3-6(8(11)13-2)10-4-5(7)9/h3-4H,1-2H3
Formel vikt 244.9688