missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromoorotate, 97%, Thermo Scientific™

Catalog Number p-7056023
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
30825-88-8
C6H5BrN2O4
249.02
MFCD07023034
UJCPEPQYSPRWGU-UHFFFAOYSA-N
methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, methyl 5-bromo-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate, methyl 5-bromo-2,6-dihydroxypyrimidine-4-carboxylate, 4-pyrimidinecarboxylic acid, 5-bromo-1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester
6324367
methyl 5-bromo-2,4-dioxo-1H-pyrimidine-6-carboxylate
COC(=O)C1=C(C(=O)NC(=O)N1)Br
This item is not returnable. View return policy
30825-88-8
C6H5BrN2O4
249.02
MFCD07023034
UJCPEPQYSPRWGU-UHFFFAOYSA-N
methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, methyl 5-bromo-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate, methyl 5-bromo-2,6-dihydroxypyrimidine-4-carboxylate, 4-pyrimidinecarboxylic acid, 5-bromo-1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester
6324367
methyl 5-bromo-2,4-dioxo-1H-pyrimidine-6-carboxylate
COC(=O)C1=C(C(=O)NC(=O)N1)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 252°C to 254°C
Quantity 250 mg
Beilstein 915036
Formula Weight 249.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 5-bromoorotate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'