Läs mer
Methyl 5-Bromovalerate, TRC
Produktkod. 30465662
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30465662 25 g
1 options
Produktkod. 30465662 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29603325G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 5-Bromovalerate
CAS 5454-83-1
Molekylformel C6H11BrO2
Kvantitet 25 g
Synonym 5-Bromopentanoic Acid Methyl Ester, 5-Bromovaleric Acid Methyl Ester, Methyl 5-Bromopentanoate, Methyl delta-Bromovalerate, Methyl ω-Bromopentanoate, NSC 23228
InChI formel InChI=1S/C6H11BrO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3
LEDER COC(=O)CCCCBr
IUPAC-namn methyl 5-bromopentanoate
Molekylvikt (g/mol) 195.05
Formel vikt 193.9942