Läs mer
Methyl 5-Chloro-3-(chlorosulfonyl)thiophene-2-carboxylate, TRC
Produktkod. 30479018
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479018 100 mg
30470550 50 mg
30483583 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479018 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM219133100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126910-68-7
Molekylformel C6H4Cl2O4S2
Molekylvikt (g/mol) 275.11
IUPAC-namn methyl 5-chloro-3-chlorosulfonylthiophene-2-carboxylate
LEDER COC(=O)c1sc(Cl)cc1S(=O)(=O)Cl
Specifikationer
Kemiskt namn eller material Methyl 5-Chloro-3-(chlorosulfonyl)thiophene-2-carboxylate
Kvantitet 100 mg
InChI formel InChI=1S/C6H4Cl2O4S2/c1-12-6(9)5-3(14(8,10)11)2-4(7)13-5/h2H,1H3
Formel vikt 273.8928