Läs mer
Methyl 5-Chloropentanoate, TRC
Produktkod. 30479696
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479696 50 mg
30476117 100 mg
30472750 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM34046050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14273-86-0
Molekylformel C6H11O2Cl
Molekylvikt (g/mol) 150.6
Synonym Apixaban Impurity 84
IUPAC-namn methyl 5-chloropentanoate
LEDER COC(=O)CCCCCl
Specifikationer
Kemiskt namn eller material Methyl 5-Chloropentanoate
Kvantitet 50 mg
InChI formel InChI=1S/C6H11ClO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3
Formel vikt 150.0448