Läs mer
methyl 5-formylfuran-2-carboxylate, TRC
Produktkod. 30470866
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470866 50 mg
30476441 250 mg
30469915 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470866 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM57136550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5904-71-2
Molekylformel C7H6O4
Molekylvikt (g/mol) 154.12
IUPAC-namn methyl 5-formylfuran-2-carboxylate
LEDER COC(=O)c1oc(C=O)cc1
Specifikationer
Kemiskt namn eller material methyl 5-formylfuran-2-carboxylate
Kvantitet 50 mg
InChI formel InChI=1S/C7H6O4/c1-10-7(9)6-3-2-5(4-8)11-6/h2-4H,1H3
Formel vikt 154.0266