Läs mer
Methyl 5-Hydroxy-2-benzimidazolecarbamate, TRC
Produktkod. 30004454
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004454 10 mg
30005683 50 mg
30011246 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31258510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22769-68-2
Molekylformel C9H9N3O3
Molekylvikt (g/mol) 207.19
Synonym 5-Hydroxy-2-methoxycarbonylaminobenzimidazole, 5-HydroxyMBC, 5-Hydroxycarbendazim, Methyl 5-hydroxybenzimidazol-2-ylcarbamate;
LEDER O=C(CC(OCC)=O)CCCOCC1=CC=CC=C1
Specifikationer
Kemiskt namn eller material Methyl 5-Hydroxy-2-benzimidazolecarbamate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
Procent renhet >95%