missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-iodosalicylate 99%, ACROS Organics™

Brand:  Acros Organics 381770500

Additional Details : CAS Number : 4068-75-1 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10026672

  • 1885.27 SEK / 50g


Call for latest availability.
+46 (0)31 352 32 30

Add to basket


Specifications

Chemical Identifiers

Methyl 5-iodosalicylate
99%
IC6H3(OH)COOCH3
MFCD00016462
≥98.5%
C8H7IO3
278.04
Methyl 2-hydroxy-5-iodobenzoate

Specifications

Methyl 5-iodosalicylate
Authentic
Crystalline Powder
99%
IC6H3(OH)COOCH3
MFCD00016462
73° to 78°C
Methyl 2-hydroxy-5-iodobenzoate
4068-75-1
Glass bottle
≥98.5%
C8H7IO3
278.04
White
50g
Videos
SDS