Läs mer
Methyl 5-Methoxy-2-methylindole-3-acetate, TRC
Produktkod. 30482843
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30482843 50 mg
1 options
Produktkod. 30482843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32048050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 5-Methoxy-2-methylindole-3-acetate
CAS 7588-36-5
Molekylformel C13 H15 N O3
Kvantitet 50 mg
Synonym Methyl 5-Methoxy-2-methylindole-3-acetate, 1H-Indole-3-acetic acid','5-methoxy-2-methyl-','methyl ester, Indole-3-acetic acid','5-methoxy-2-methyl-','methyl ester (7CI,8CI), 5-Methoxy-2-methylindole-3-acetic acid methyl ester, Methyl 5-methoxy-2-methylindole-3-acetate
InChI formel InChI=1S/C13H15NO3/c1-8-10(7-13(15)17-3)11-6-9(16-2)4-5-12(11)14-8/h4-6,14H,7H2,1-3H3
LEDER COC(=O)Cc1c(C)[nH]c2ccc(OC)cc12
IUPAC-namn methyl 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate
Molekylvikt (g/mol) 233.26
Formel vikt 233.1052