Läs mer
Methyl 5-Nitroisophthalate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-Nitro-isophthalic acid monomethyl ester |
| CAS | 1955-46-0 |
| Molekylformel | C9 H7 N O6 |
| Kvantitet | 50 g |
| Synonym | 5-Nitro-1,3-benzenedicarboxylic Acid 1-Methyl Ester, 5-Nitro-1,3-benzenedicarboxylic Acid Monomethyl Ester, 5-Nitro-Isophthalic Acid Methyl Ester, 5-Nitro-Isophthalic Acid Monomethyl Ester, 3-Carbomethoxy-5-nitrobenzoic Acid, 3-Methoxycarbonyl-5-nitrobenzoic Acid, 3-Nitro-5-(methoxycarbonyl)benzoic Acid, 5-Nitro-1,3-benzenedicarboxylic Acid Methyl Ester, 5-Nitro-1,3-benzenedicarboxylic Acid Monomethyl Ester, 5-Nitro-3-(carbomethoxy)benzoic Acid, 5-Nitroisophthalic Acid Methyl Ester, 5-Nitroisophthalic Acid Monomethyl Ester, Methyl 3-carboxy-5-nitrobenzoate, Methyl Hydrogen 5-Nitroisophthalate, Monomethyl 5-Nitroisophthalate |
| InChI formel | InChI=1S/C9H7NO6/c1-16-9(13)6-2-5(8(11)12)3-7(4-6)10(14)15/h2-4H,1H3,(H,11,12) |
| LEDER | COC(=O)c1cc(cc(c1)[N+](=O)[O-])C(=O)O |
| IUPAC-namn | 3-methoxycarbonyl-5-nitrobenzoic acid |
| Molekylvikt (g/mol) | 225.16 |
| Formel vikt | 225.0273 |