missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Methyl (5Beta,6Alpha)-6-Ethyl-3,7-dioxo-cholan-24-oic Acid Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl (5beta,6alpha)-6-Ethyl-3,7-dioxo-cholan-24-oic Acid Ester |
| CAS | 1926987-00-9 |
| Molekylformel | C27H42O4 |
| Kvantitet | 5 mg |
| InChI formel | InChI=1S/C27H42O4/c1-6-18-22-15-17(28)11-13-27(22,4)21-12-14-26(3)19(16(2)7-10-23(29)31-5)8-9-20(26)24(21)25(18)30/h16,18-22,24H,6-15H2,1-5H3/t16-,18-,19-,20+,21+,22+,24+,26-,27-/m1/s1 |
| LEDER | CC[C@@H]1[C@@H]2CC(=O)CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3C1=O)[C@H](C)CCC(=O)OC |
| IUPAC-namn | methyl (4R)-4-[(5S,6R,8S,9S,10S,13R,14S,17R)-6-ethyl-10,13-dimethyl-3,7-dioxo-2,4,5,6,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate |
| Molekylvikt (g/mol) | 430.62 |
| Formel vikt | 430.3083 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?