Läs mer
Methyl 6-[[[(2-Chloroethyl)amino]carbonyl]amino]-6-deoxy-Alpha-D-glucopyranoside, TRC
Produktkod. 30472371
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30472371 25 mg
1 options
Produktkod. 30472371 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30442025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 6-[(((2-Chloroethyl)amino)carbonyl)amino]-6-deoxy-Alpha-D-glucopyranoside
CAS 58994-95-9
Molekylformel C10H19ClN2O6
Kvantitet 25 mg
Synonym 1-(2-Chloroethyl)-3-(((2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxytetrahydro-2H-pyran-2-yl)methyl)urea
InChI formel InChI=1S/C10H19ClN2O6/c1-18-9-8(16)7(15)6(14)5(19-9)4-13-10(17)12-3-2-11/h5-9,14-16H,2-4H2,1H3,(H2,12,13,17)/t5-,6-,7+,8-,9+/m1/s1
LEDER CO[C@H]1O[C@H](CNC(=O)NCCCl)[C@@H](O)[C@H](O)[C@H]1O
Molekylvikt (g/mol) 298.72