Läs mer
Methyl 6-methylpyridine-2-carboxylate, 96%, Thermo Scientific Chemicals
Produktkod. 15406407
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
Produktkod. 15406407
Brand: Thermo Scientific Alfa Aesar H33571.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13602-11-4
Molekylformel C8H9NO2
Molekylvikt (g/mol) 151.17
MDL-nummer MFCD06203670
InChI-nyckel CYWIZMOZBTXFIL-UHFFFAOYSA-N
Synonym methyl 6-methylpicolinate, 6-methylpyridine-2-carboxylic acid methyl ester, methyl 6-methyl-2-pyridinecarboxylate, 6-methyl-pyridine-2-carboxylic acid methyl ester, 6-methylpicolinic acid methyl ester, 2-pyridinecarboxylic acid, 6-methyl-, methyl ester, pubchem20955, methyl-6-methylpicolinate, ksc495e0b, aronis24547
PubChem CID 73022
IUPAC-namn methyl 6-methylpyridine-2-carboxylate
LEDER COC(=O)C1=CC=CC(C)=N1
Specifikationer
Kemiskt namn eller material Methyl 6-methylpyridine-2-carboxylate
Kokpunkt 242°C
Kvantitet 250 mg
Formel vikt 151.17
Procent renhet 96%